RIE156325261 Rieke Metals, Inc.

2-pyrrolidinomethyl-4'-thiomethylbenzophenone

  • MDL number MFCD03842530
  • Cas Number : 898774-41-9
  • Molecular Weight : 311.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0300-15

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0300-15
Availability details for
Quantity
  • Availability
  • Add to Cart