RIE156325262 Rieke Metals, Inc.

3'-bromo-2-pyrrolidinomethyl benzophenone

  • MDL number MFCD03842531
  • Cas Number : 898774-44-2
  • Molecular Weight : 344.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0300-18

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0300-18
Availability details for
Quantity
  • Availability
  • Add to Cart