RIE156325266 Rieke Metals, Inc.

4-methoxy-4'-pyrrolidinomethyl benzophenone

  • MDL number MFCD03841501
  • Cas Number : 898776-01-7
  • Molecular Weight : 295.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6124-0300-07

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6124-0300-07
Availability details for
Quantity
  • Availability
  • Add to Cart