RIE156326245 Rieke Metals, Inc.

3',4'-dimethyl-2-pyrrolidinomethyl benzophenone

  • MDL number MFCD03842541
  • Cas Number : 898774-65-7
  • Molecular Weight : 293.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3,4-dimethylphenyl)[2-(1-pyrrolidinylmethyl)phenyl]methanone
Inchi Code 1S/C20H23NO/c1-15-9-10-17(13-16(15)2)20(22)19-8-4-3-7-18(19)14-21-11-5-6-12-21/h3-4,7-10,13H,5-6,11-12,14H2,1-2H3
Inchi Key RENUCQNPLAHLNQ-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6126-0300-30

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0300-30
Availability details for
Quantity
  • Availability
  • Add to Cart