RIE156326246 Rieke Metals, Inc.

3',5'-dimethyl-2-pyrrolidinomethyl benzophenone

  • MDL number MFCD03842542
  • Cas Number : 898774-67-9
  • Molecular Weight : 293.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0300-31

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0300-31
Availability details for
Quantity
  • Availability
  • Add to Cart