RIE156326249 Rieke Metals, Inc.

3'-chloro-4'-fluoro-2-pyrrolidinomethyl benzophenone

  • MDL number MFCD03842545
  • Cas Number : 898774-73-7
  • Molecular Weight : 317.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0300-34

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0300-34
Availability details for
Quantity
  • Availability
  • Add to Cart