RIE156326250 Rieke Metals, Inc.

2-chloro-2'-pyrrolidinomethyl benzophenone

  • MDL number MFCD03842546
  • Cas Number : 898774-75-9
  • Molecular Weight : 299.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2-chlorophenyl)[2-(1-pyrrolidinylmethyl)phenyl]methanone
Inchi Code 1S/C18H18ClNO/c19-17-10-4-3-9-16(17)18(21)15-8-2-1-7-14(15)13-20-11-5-6-12-20/h1-4,7-10H,5-6,11-13H2
Inchi Key QRKDAYZDYXHLLF-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6126-0300-36

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0300-36
Availability details for
Quantity
  • Availability
  • Add to Cart