RIE156326252 Rieke Metals, Inc.

2-pyrrolidinomethyl-2'-trifluoromethylbenzophenone

  • MDL number MFCD03842548
  • Cas Number : 898774-79-3
  • Molecular Weight : 333.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name [2-(1-pyrrolidinylmethyl)phenyl][2-(trifluoromethyl)phenyl]methanone
Inchi Code 1S/C19H18F3NO/c20-19(21,22)17-10-4-3-9-16(17)18(24)15-8-2-1-7-14(15)13-23-11-5-6-12-23/h1-4,7-10H,5-6,11-13H2
Inchi Key QBVDOAQREOUYIN-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6126-0300-43

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0300-43
Availability details for
Quantity
  • Availability
  • Add to Cart