RIE156326253 Rieke Metals, Inc.

2-pyrrolidinomethyl-3'-trifluoromethylbenzophenone

  • MDL number MFCD03842549
  • Cas Number : 898774-81-7
  • Molecular Weight : 333.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0300-44

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0300-44
Availability details for
Quantity
  • Availability
  • Add to Cart