RIE156326255 Rieke Metals, Inc.

4-bromo-2-fluoro-2'-pyrrolidinomethyl benzophenone

  • MDL number MFCD03842551
  • Cas Number : 898774-85-1
  • Molecular Weight : 362.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0300-49

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0300-49
Availability details for
Quantity
  • Availability
  • Add to Cart