RIE156326256 Rieke Metals, Inc.

2-chloro-4-fluoro-2'-pyrrolidinomethyl benzophenone

  • MDL number MFCD03842552
  • Cas Number : 898774-87-3
  • Molecular Weight : 317.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0300-50

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0300-50
Availability details for
Quantity
  • Availability
  • Add to Cart