RIE156326266 Rieke Metals, Inc.

3,5-difluoro-2'-pyrrolidinomethyl benzophenone

  • MDL number MFCD03842562
  • Cas Number : 898775-09-2
  • Molecular Weight : 301.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3,5-difluorophenyl)[2-(1-pyrrolidinylmethyl)phenyl]methanone
Inchi Code 1S/C18H17F2NO/c19-15-9-14(10-16(20)11-15)18(22)17-6-2-1-5-13(17)12-21-7-3-4-8-21/h1-2,5-6,9-11H,3-4,7-8,12H2
Inchi Key XUKVWLMOYONVKL-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6126-0300-60

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0300-60
Availability details for
Quantity
  • Availability
  • Add to Cart