RIE156326267 Rieke Metals, Inc.

2'-pyrrolidinomethyl-3,4,5-trifluorobenzophenone

  • MDL number MFCD03842563
  • Cas Number : 898775-12-7
  • Molecular Weight : 319.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0300-61

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0300-61
Availability details for
Quantity
  • Availability
  • Add to Cart