RIE156326268 Rieke Metals, Inc.

cyclopropyl 2-(pyrrolidinomethyl)phenyl ketone

  • MDL number MFCD03842564
  • Cas Number : 898775-15-0
  • Molecular Weight : 229.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name cyclopropyl[2-(1-pyrrolidinylmethyl)phenyl]methanone
Inchi Code 1S/C15H19NO/c17-15(12-7-8-12)14-6-2-1-5-13(14)11-16-9-3-4-10-16/h1-2,5-6,12H,3-4,7-11H2
Inchi Key HHQWGBNVTNPUQP-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6126-0301-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0301-00
Availability details for
Quantity
  • Availability
  • Add to Cart