RIE156326269 Rieke Metals, Inc.

cyclobutyl 2-(pyrrolidinomethyl)phenyl ketone

  • MDL number MFCD03842565
  • Cas Number : 898775-18-3
  • Molecular Weight : 243.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name cyclobutyl[2-(1-pyrrolidinylmethyl)phenyl]methanone
Inchi Code 1S/C16H21NO/c18-16(13-7-5-8-13)15-9-2-1-6-14(15)12-17-10-3-4-11-17/h1-2,6,9,13H,3-5,7-8,10-12H2
Inchi Key VOJYXYVOZKXUKP-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6126-0302-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0302-00
Availability details for
Quantity
  • Availability
  • Add to Cart