RIE156326270 Rieke Metals, Inc.

cyclopentyl 2-(pyrrolidinomethyl)phenyl ketone

  • MDL number MFCD03842566
  • Cas Number : 898775-21-8
  • Molecular Weight : 257.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name cyclopentyl[2-(1-pyrrolidinylmethyl)phenyl]methanone
Inchi Code 1S/C17H23NO/c19-17(14-7-1-2-8-14)16-10-4-3-9-15(16)13-18-11-5-6-12-18/h3-4,9-10,14H,1-2,5-8,11-13H2
Inchi Key ONLFCBAEGHKDEM-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6126-0303-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0303-00
Availability details for
Quantity
  • Availability
  • Add to Cart