RIE156326272 Rieke Metals, Inc.

ethyl 4-oxo-4-[2-(pyrrolidinomethyl)phenyl]butyrate

  • MDL number MFCD03842568
  • Cas Number : 898775-27-4
  • Molecular Weight : 289.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0305-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0305-00
Availability details for
Quantity
  • Availability
  • Add to Cart