RIE156326273 Rieke Metals, Inc.

ethyl 5-oxo-5-[2-(pyrrolidinomethyl)phenyl]valerate

  • MDL number MFCD03842569
  • Cas Number : 898775-30-9
  • Molecular Weight : 303.40

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0306-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0306-00
Availability details for
Quantity
  • Availability
  • Add to Cart