RIE156326274 Rieke Metals, Inc.

ethyl 6-oxo-6-[2-(pyrrolidinomethyl)phenyl]hexanoate

  • MDL number MFCD03842570
  • Cas Number : 898775-33-2
  • Molecular Weight : 317.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 6-oxo-6-[2-(1-pyrrolidinylmethyl)phenyl]hexanoate
Inchi Code 1S/C19H27NO3/c1-2-23-19(22)12-6-5-11-18(21)17-10-4-3-9-16(17)15-20-13-7-8-14-20/h3-4,9-10H,2,5-8,11-15H2,1H3
Inchi Key DRTCWRPRZXAXCK-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6126-0307-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0307-00
Availability details for
Quantity
  • Availability
  • Add to Cart