RIE156326337 Rieke Metals, Inc.

2-[8-(1,4-dioxa-8-azaspiro[4.5]decyl)methyl]-4'-methyl benzophenone

  • MDL number MFCD03842690
  • Cas Number : 898755-77-6
  • Molecular Weight : 351.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0600-04

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0600-04
Availability details for
Quantity
  • Availability
  • Add to Cart