RIE156326339 Rieke Metals, Inc.

2-[8-(1,4-dioxa-8-azaspiro[4.5]decyl)methyl]-3'-methoxy benzophenone

  • MDL number MFCD03842692
  • Cas Number : 898755-80-1
  • Molecular Weight : 367.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0600-06

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0600-06
Availability details for
Quantity
  • Availability
  • Add to Cart