RIE156326343 Rieke Metals, Inc.

4'-cyano-2-[8-(1,4-dioxa-8-azaspiro[4.5]decyl)methyl]benzophenone

  • MDL number MFCD03842696
  • Cas Number : 898755-87-8
  • Molecular Weight : 362.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0600-10

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0600-10
Availability details for
Quantity
  • Availability
  • Add to Cart