RIE156326347 Rieke Metals, Inc.

2-[8-(1,4-dioxa-8-azaspiro[4.5]decyl)methyl]-2'-thiomethyl benzophenone

  • MDL number MFCD03842700
  • Cas Number : 898755-94-7
  • Molecular Weight : 383.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0600-14

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0600-14
Availability details for
Quantity
  • Availability
  • Add to Cart