RIE156326350 Rieke Metals, Inc.

4'-bromo-2-[8-(1,4-dioxa-8-azaspiro[4.5]decyl)methyl]benzophenone

  • MDL number MFCD03842703
  • Cas Number : 898755-99-2
  • Molecular Weight : 416.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-bromophenyl)[2-(1,4-dioxa-8-azaspiro[4.5]dec-8-ylmethyl)phenyl]methanone
Inchi Code 1S/C21H22BrNO3/c22-18-7-5-16(6-8-18)20(24)19-4-2-1-3-17(19)15-23-11-9-21(10-12-23)25-13-14-26-21/h1-8H,9-15H2
Inchi Key JDWZLXAWRDRFFI-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6126-0600-19

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0600-19
Availability details for
Quantity
  • Availability
  • Add to Cart