RIE156326351 Rieke Metals, Inc.

3'-chloro-2-[8-(1,4-dioxa-8-azaspiro[4.5]decyl)methyl]benozphenone

  • MDL number MFCD03842704
  • Cas Number : 898756-00-8
  • Molecular Weight : 371.87

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0600-20

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0600-20
Availability details for
Quantity
  • Availability
  • Add to Cart