RIE156326353 Rieke Metals, Inc.

2-[8-(1,4-dioxa-8-azaspiro[4.5]decyl)methyl]-3'-fluorobenzophenone

  • MDL number MFCD03842706
  • Cas Number : 898756-04-2
  • Molecular Weight : 355.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name [2-(1,4-dioxa-8-azaspiro[4.5]dec-8-ylmethyl)phenyl](3-fluorophenyl)methanone
Inchi Code 1S/C21H22FNO3/c22-18-6-3-5-16(14-18)20(24)19-7-2-1-4-17(19)15-23-10-8-21(9-11-23)25-12-13-26-21/h1-7,14H,8-13,15H2
Inchi Key BPEMXCIJWBBQKO-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6126-0600-22

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0600-22
Availability details for
Quantity
  • Availability
  • Add to Cart