RIE156326354 Rieke Metals, Inc.

2-[8-(1,4-dioxa-8-azaspiro[4.5]decyl)methyl]-4'-fluorobenzophenone

  • MDL number MFCD03842707
  • Cas Number : 898756-05-3
  • Molecular Weight : 355.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0600-23

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0600-23
Availability details for
Quantity
  • Availability
  • Add to Cart