RIE156326358 Rieke Metals, Inc.

2,6-dimethyl-2'-[8-(1,4-dioxa-8-azaspiro[4.5]decyl)methyl]benzophenone

  • MDL number MFCD03842711
  • Cas Number : 898756-12-2
  • Molecular Weight : 365.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0600-29

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0600-29
Availability details for
Quantity
  • Availability
  • Add to Cart