RIE156326359 Rieke Metals, Inc.

3',4'-dimethyl-2-[8-(1,4-dioxa-8-azaspiro[4.5]decyl)methyl]benzophenone

  • MDL number MFCD03842712
  • Cas Number : 898756-14-4
  • Molecular Weight : 365.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3,4-dimethylphenyl)[2-(1,4-dioxa-8-azaspiro[4.5]dec-8-ylmethyl)phenyl]methanone
Inchi Code 1S/C23H27NO3/c1-17-7-8-19(15-18(17)2)22(25)21-6-4-3-5-20(21)16-24-11-9-23(10-12-24)26-13-14-27-23/h3-8,15H,9-14,16H2,1-2H3
Inchi Key ITEJEHBFGWBFIH-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6126-0600-30

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0600-30
Availability details for
Quantity
  • Availability
  • Add to Cart