RIE156326360 Rieke Metals, Inc.

3',5'-dimethyl-2-[8-(1,4-dioxa-8-azaspiro[4.5]decyl)methyl]benzophenone

  • MDL number MFCD03842713
  • Cas Number : 898756-15-5
  • Molecular Weight : 365.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3,5-dimethylphenyl)[2-(1,4-dioxa-8-azaspiro[4.5]dec-8-ylmethyl)phenyl]methanone
Inchi Code 1S/C23H27NO3/c1-17-13-18(2)15-20(14-17)22(25)21-6-4-3-5-19(21)16-24-9-7-23(8-10-24)26-11-12-27-23/h3-6,13-15H,7-12,16H2,1-2H3
Inchi Key JXGZWSAYXCHCKW-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6126-0600-31

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0600-31
Availability details for
Quantity
  • Availability
  • Add to Cart