RIE156326361 Rieke Metals, Inc.

4-bromo-2'-[8-(1,4-dioxa-8-azaspiro[4.5]decyl)methyl]-3-fluorobenzophenone

  • MDL number MFCD07699570
  • Cas Number : 898756-17-7
  • Molecular Weight : 434.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-bromo-3-fluorophenyl)[2-(1,4-dioxa-8-azaspiro[4.5]dec-8-ylmethyl)phenyl]methanone
Inchi Code 1S/C21H21BrFNO3/c22-18-6-5-15(13-19(18)23)20(25)17-4-2-1-3-16(17)14-24-9-7-21(8-10-24)26-11-12-27-21/h1-6,13H,7-12,14H2
Inchi Key NYXXDXWYQJWBDC-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6126-0600-32

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0600-32
Availability details for
Quantity
  • Availability
  • Add to Cart