RIE156326372 Rieke Metals, Inc.

4-chloro-2'-[8-(1,4-dioxa-8-azaspiro[4.5]decyl)methyl]-2-fluorobenzophenone

  • MDL number MFCD03842724
  • Cas Number : 898756-45-1
  • Molecular Weight : 389.85

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-chloro-2-fluorophenyl)[2-(1,4-dioxa-8-azaspiro[4.5]dec-8-ylmethyl)phenyl]methanone
Inchi Code 1S/C21H21ClFNO3/c22-16-5-6-18(19(23)13-16)20(25)17-4-2-1-3-15(17)14-24-9-7-21(8-10-24)26-11-12-27-21/h1-6,13H,7-12,14H2
Inchi Key NYWSZPXBSGHRFL-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6126-0600-52

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0600-52
Availability details for
Quantity
  • Availability
  • Add to Cart