RIE156326379 Rieke Metals, Inc.

3,4-difluoro-2'-[8-(1,4-dioxa-8-azaspiro[4.5]decyl)methyl]benzophenone

  • MDL number MFCD03842731
  • Cas Number : 898756-66-6
  • Molecular Weight : 373.40

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0600-59

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0600-59
Availability details for
Quantity
  • Availability
  • Add to Cart