RIE156326384 Rieke Metals, Inc.

cyclopentyl 2-[8-(1,4-dioxa-8-azaspiro[4.5]decyl)methyl]phenyl ketone

  • MDL number MFCD03842736
  • Cas Number : 898781-17-4
  • Molecular Weight : 329.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0603-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0603-00
Availability details for
Quantity
  • Availability
  • Add to Cart