RIE156326388 Rieke Metals, Inc.

ethyl 6-[2-[8-(1,4-dioxa-8-azaspiro[4.5]decyl)methyl]phenyl]-6-oxohexanoate

  • MDL number MFCD03842740
  • Cas Number : 898781-29-8
  • Molecular Weight : 389.50

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0607-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0607-00
Availability details for
Quantity
  • Availability
  • Add to Cart