RIE156326391 Rieke Metals, Inc.

2-(thiomorpholinomethyl)benzophenone

  • MDL number MFCD03842743
  • Cas Number : 745720-16-5
  • Molecular Weight : 297.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name phenyl[2-(4-thiomorpholinylmethyl)phenyl]methanone
Inchi Code 1S/C18H19NOS/c20-18(15-6-2-1-3-7-15)17-9-5-4-8-16(17)14-19-10-12-21-13-11-19/h1-9H,10-14H2
Inchi Key OLRDMSJFFYWEGS-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6126-0700-01

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0700-01
Availability details for
Quantity
  • Availability
  • Add to Cart