RIE156326392 Rieke Metals, Inc.

2-methyl-2'-thiomorpholinomethyl benzophenone

  • MDL number MFCD03842744
  • Cas Number : 898781-38-9
  • Molecular Weight : 311.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0700-02

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0700-02
Availability details for
Quantity
  • Availability
  • Add to Cart