RIE156326399 Rieke Metals, Inc.

3'-cyano-2-thiomorpholinomethyl benzophenone

  • MDL number MFCD03842751
  • Cas Number : 898781-56-1
  • Molecular Weight : 322.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0700-09

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0700-09
Availability details for
Quantity
  • Availability
  • Add to Cart