RIE156326400 Rieke Metals, Inc.

4'-cyano-2-thiomorpholinomethyl benzophenone

  • MDL number MFCD03842752
  • Cas Number : 898781-58-3
  • Molecular Weight : 322.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-[2-(4-thiomorpholinylmethyl)benzoyl]benzonitrile
Inchi Code 1S/C19H18N2OS/c20-13-15-5-7-16(8-6-15)19(22)18-4-2-1-3-17(18)14-21-9-11-23-12-10-21/h1-8H,9-12,14H2
Inchi Key UTABLOYIIQKJKF-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6126-0700-10

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0700-10
Availability details for
Quantity
  • Availability
  • Add to Cart