RIE156326403 Rieke Metals, Inc.

4'-carboethoxy-2-thiomorpholinomethyl benzophenone

  • MDL number MFCD03842755
  • Cas Number : 898781-64-1
  • Molecular Weight : 369.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 4-[2-(4-thiomorpholinylmethyl)benzoyl]benzoate
Inchi Code 1S/C21H23NO3S/c1-2-25-21(24)17-9-7-16(8-10-17)20(23)19-6-4-3-5-18(19)15-22-11-13-26-14-12-22/h3-10H,2,11-15H2,1H3
Inchi Key CHNTVUOTRHCYRP-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6126-0700-13

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0700-13
Availability details for
Quantity
  • Availability
  • Add to Cart