RIE156326405 Rieke Metals, Inc.

4'-thiomethyl-2-thiomorpholinomethyl benzophenone

  • MDL number MFCD03842757
  • Cas Number : 898781-68-5
  • Molecular Weight : 343.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0700-15

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0700-15
Availability details for
Quantity
  • Availability
  • Add to Cart