RIE156326407 Rieke Metals, Inc.

4'-bromo-2-thiomorpholinomethyl benzophenone

  • MDL number MFCD03842759
  • Cas Number : 898781-72-1
  • Molecular Weight : 376.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0700-19

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0700-19
Availability details for
Quantity
  • Availability
  • Add to Cart