RIE156326413 Rieke Metals, Inc.

2,4-dimethyl-2'-thiomorpholinomethyl benzophenone

  • MDL number MFCD03842765
  • Cas Number : 898781-84-5
  • Molecular Weight : 325.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0700-27

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0700-27
Availability details for
Quantity
  • Availability
  • Add to Cart