RIE156326422 Rieke Metals, Inc.

2-fluoro-2'-thiomorpholinomethyl benzophenone

  • MDL number MFCD03842774
  • Cas Number : 898782-02-0
  • Molecular Weight : 315.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0700-37

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0700-37
Availability details for
Quantity
  • Availability
  • Add to Cart