RIE156326424 Rieke Metals, Inc.

2-thiomorpholinomethyl-3'-trifluoromethylbenzophenone

  • MDL number MFCD03842776
  • Cas Number : 898782-08-6
  • Molecular Weight : 365.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0700-44

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0700-44
Availability details for
Quantity
  • Availability
  • Add to Cart