RIE156326426 Rieke Metals, Inc.

4-bromo-2-fluoro-2'-thiomorpholinomethyl benzophenone

  • MDL number MFCD03842778
  • Cas Number : 898782-14-4
  • Molecular Weight : 394.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-bromo-2-fluorophenyl)[2-(4-thiomorpholinylmethyl)phenyl]methanone
Inchi Code 1S/C18H17BrFNOS/c19-14-5-6-16(17(20)11-14)18(22)15-4-2-1-3-13(15)12-21-7-9-23-10-8-21/h1-6,11H,7-10,12H2
Inchi Key ARKVTRAABJFJMB-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6126-0700-49

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0700-49
Availability details for
Quantity
  • Availability
  • Add to Cart