RIE156326430 Rieke Metals, Inc.

2,3-dichloro-2'-thiomorpholinomethyl benzophenone

  • MDL number MFCD03842782
  • Cas Number : 898782-26-8
  • Molecular Weight : 366.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0700-53

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0700-53
Availability details for
Quantity
  • Availability
  • Add to Cart