RIE156326433 Rieke Metals, Inc.

3,4-dichloro-2'-thiomorpholinomethyl benzophenone

  • MDL number MFCD03842785
  • Cas Number : 898782-35-9
  • Molecular Weight : 366.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0700-56

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0700-56
Availability details for
Quantity
  • Availability
  • Add to Cart