RIE156326439 Rieke Metals, Inc.

cyclopropyl 2-(thiomorpholinomethyl)phenyl ketone

  • MDL number MFCD03842791
  • Cas Number : 898782-52-0
  • Molecular Weight : 261.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name cyclopropyl[2-(4-thiomorpholinylmethyl)phenyl]methanone
Inchi Code 1S/C15H19NOS/c17-15(12-5-6-12)14-4-2-1-3-13(14)11-16-7-9-18-10-8-16/h1-4,12H,5-11H2
Inchi Key ADVSKYIHIKPPIQ-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6126-0701-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0701-00
Availability details for
Quantity
  • Availability
  • Add to Cart