RIE156326440 Rieke Metals, Inc.

cyclobutyl 2-(thiomorpholinomethyl)phenyl ketone

  • MDL number MFCD03842792
  • Cas Number : 898782-54-2
  • Molecular Weight : 275.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6126-0702-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6126-0702-00
Availability details for
Quantity
  • Availability
  • Add to Cart